KEY001132698 Key Organics / BIONET

N-(2-hydroxyethyl)-2-[(4-methoxyphenyl)formamido]acetamide

  • MDL number MFCD00140894
  • Cas Number : 338396-48-8
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148 - 150
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67006

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2F-906_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart