KEY001132726 Key Organics / BIONET

ethyl 2-amino-4-phenyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD00171776
  • Cas Number : 147353-77-3
  • Molecular Weight : 230.267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157 - 158
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67046

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2F-958_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart