KEY001132823 Key Organics / BIONET

N-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-3-(hydrazinecarbonyl)-5-methyl-1,2-oxazole-4-carboxamide

  • MDL number MFCD00664740
  • Cas Number : 338397-37-8
  • Molecular Weight : 363.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146 - 148
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67197

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2G-939_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart