KEY001132867 Key Organics / BIONET

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-pyrrole-2-sulfonyl chloride

  • MDL number MFCD00214861
  • Cas Number : 338397-79-8
  • Molecular Weight : 345.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 71 - 74
Iupac Name 1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-1H-pyrrole-2-sulfonyl chloride
Inchi Code 1S/C10H5Cl2F3N2O2S/c11-7-4-6(10(13,14)15)5-16-9(7)17-3-1-2-8(17)20(12,18)19/h1-5H
Inchi Key YHIKQCOKDZJLOK-UHFFFAOYSA-N

Description

Melting Point 71 - 74
Purity 95%
Storage temp. Store under N2 at 4
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67243

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/2H-054_SDS.pdf
hcodes H302,H312,H314,H315,H319,H332,H335
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart