KEY001132879 Key Organics / BIONET

2,2,2-trichloro-1-{4-[2-(4-chlorophenyl)acetyl]-1H-pyrrol-2-yl}ethan-1-one

  • MDL number MFCD00173091
  • Cas Number : 338397-93-6
  • Molecular Weight : 365.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147 - 150
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67257

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2H-070_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart