KEY001132883 Key Organics / BIONET

4-[2-(4-chlorophenyl)acetyl]-1H-pyrrole-2-carboxamide

  • MDL number MFCD00214866
  • Cas Number : 338397-97-0
  • Molecular Weight : 262.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260 - 263
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67261

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2H-074_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart