KEY001132978 Key Organics / BIONET

5-methyl-2-(pyridin-4-yl)-1,3-thiazol-4-ol

  • MDL number MFCD00172519
  • Cas Number : 70547-50-1
  • Molecular Weight : 192.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 221 - 225
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67377

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2H-451S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart