KEY001132997 Key Organics / BIONET

5-methyl-2-(pyridin-2-yl)-1,3-thiazol-4-ol

  • MDL number MFCD00172537
  • Cas Number : 131786-47-5
  • Molecular Weight : 192.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235 - 238
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67403

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2H-480S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart