KEY001133014 Key Organics / BIONET

1-[(4-chlorophenyl)methyl]-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

  • MDL number MFCD00172960
  • Cas Number : 338399-17-0
  • Molecular Weight : 261.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210 - 212
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67424

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2H-907_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart