KEY001133080 Key Organics / BIONET

4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}butanoic acid

  • MDL number MFCD00172556
  • Cas Number : 338770-18-6
  • Molecular Weight : 282.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92 - 95
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67494

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2H-982_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart