KEY001133085 Key Organics / BIONET

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1H-indole-3-carbaldehyde

  • MDL number MFCD00664496
  • Cas Number : 338770-32-4
  • Molecular Weight : 324.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132 - 134
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67499

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2H-989_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart