KEY001133544 Key Organics / BIONET

3-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]sulfanyl}-5-methyl-1,2-thiazole-4-carbonitrile

  • MDL number MFCD00244313
  • Cas Number : 338775-63-6
  • Molecular Weight : 335.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122 - 125
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68017

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2L-051_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart