KEY001133669 Key Organics / BIONET

2,4-dioxo-1-(prop-2-en-1-yl)-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

  • MDL number MFCD01315928
  • Cas Number : 25855-27-0
  • Molecular Weight : 177.163

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156 - 158
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68154

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2L-903_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart