KEY001133671 Key Organics / BIONET

4-chloro-2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5-[(2-hydroxyethyl)(methyl)amino]-2,3-dihydropyridazin-3-one

  • MDL number MFCD00793844
  • Cas Number : 338399-91-0
  • Molecular Weight : 383.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149 - 151
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68156

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2L-905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart