KEY001133832 Key Organics / BIONET

ethyl [(2-benzoylphenyl)carbamoyl]formate

  • MDL number MFCD00974203
  • Cas Number : 338401-00-6
  • Molecular Weight : 297.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122 - 124
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68341

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2M-907_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart