KEY001133848 Key Organics / BIONET

3-phenyl-1H-indole-2-carboxylic acid

  • MDL number MFCD00974212
  • Cas Number : 6915-67-9
  • Molecular Weight : 237.258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195 - 197
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68356

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2M-923_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart