KEY001134016 Key Organics / BIONET

(E)-N-{[2-(3-chlorophenoxy)phenyl]methylidene}hydroxylamine

  • MDL number MFCD02091012
  • Cas Number : 449778-76-1
  • Molecular Weight : 247.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85 - 87
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68552

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart