KEY001134022 Key Organics / BIONET

(E)-N-{[2-(4-fluorophenoxy)phenyl]methylidene}hydroxylamine

  • MDL number MFCD02091015
  • Cas Number : 449778-79-4
  • Molecular Weight : 231.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 107 - 109
Iupac Name 2-(4-fluorophenoxy)benzaldehyde oxime
Inchi Code 1S/C13H10FNO2/c14-11-5-7-12(8-6-11)17-13-4-2-1-3-10(13)9-15-16/h1-8H,9H2
Inchi Key NIQIVSWDDCYLCC-UHFFFAOYSA-N

Description

Melting Point 107 - 109
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68558

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/2P-011_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart