KEY001134057 Key Organics / BIONET

{2-[4-(benzyloxy)phenoxy]phenyl}methanol

  • MDL number MFCD01814787
  • Cas Number : 449778-82-9
  • Molecular Weight : 306.361

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75 - 77
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68614

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-080_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart