KEY001134387 Key Organics / BIONET

4,6-dimethyl-1H-pyrazolo[3,4-b]pyridin-3-amine

  • MDL number MFCD00124667
  • Cas Number : 41601-44-9
  • Molecular Weight : 162.196

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 273 - 274
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68986

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2R-0620_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart