KEY001134426 Key Organics / BIONET

2-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)benzoic acid

  • MDL number MFCD02186096
  • Cas Number : 4770-69-8
  • Molecular Weight : 253.257

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240 - 243
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69036

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2R-0689_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart