KEY001134741 Key Organics / BIONET

4-[(1,3,5-trimethyl-1H-pyrazol-4-yl)methyl]phenol

  • MDL number MFCD00138770
  • Cas Number : 75999-00-7
  • Molecular Weight : 216.284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170 - 171
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70172

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3C-113_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart