KEY001134796 Key Organics / BIONET

3-{[(2,6-dichlorophenyl)methyl]sulfanyl}-1H-1,2,4-triazol-5-amine

  • MDL number MFCD00138373
  • Cas Number : 261928-97-6
  • Molecular Weight : 275.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70226

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3D-079_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart