KEY001134931 Key Organics / BIONET

methyl 3-benzamido-6-methyl-2-oxo-2H-pyran-5-carboxylate

  • MDL number MFCD00139779
  • Cas Number : 338404-89-0
  • Molecular Weight : 287.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170 - 173
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70388

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3E-925_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart