KEY001134968 Key Organics / BIONET

1-(hydrazinecarbonyl)-N-[4-(trifluoromethoxy)phenyl]formamide

  • MDL number MFCD00173093
  • Cas Number : 338405-24-6
  • Molecular Weight : 263.176

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215 - 217
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70431

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3F-035_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart