KEY001134984 Key Organics / BIONET

1-(hydrazinecarbonyl)-N-phenylformamide

  • MDL number MFCD00173094
  • Cas Number : 4740-46-9
  • Molecular Weight : 179.179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215 - 217
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70448

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3F-053_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart