KEY001135076 Key Organics / BIONET

methyl 2-{[4-formyl-1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]sulfanyl}benzoate

  • MDL number MFCD00140853
  • Cas Number : 321553-50-8
  • Molecular Weight : 344.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158 - 160
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70556

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3F-384S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart