KEY001135156 Key Organics / BIONET

2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2-(4-chlorophenyl)acetonitrile

  • MDL number MFCD00172211
  • Cas Number : 213993-80-7
  • Molecular Weight : 331.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47 - 49 / 146@0.1mm Hg
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70645

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3G-030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart