KEY001135158 Key Organics / BIONET

2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2-(4-methoxyphenyl)acetonitrile

  • MDL number MFCD00172212
  • Cas Number : 338407-10-6
  • Molecular Weight : 326.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100 - 102
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70647

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3G-032_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart