KEY001135260 Key Organics / BIONET

ethyl 4-(4-bromobenzoyl)-1,2-oxazole-3-carboxylate

  • MDL number MFCD00664751
  • Cas Number : 338408-83-6
  • Molecular Weight : 324.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78 - 80
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70802

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3G-905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart