KEY001135263 Key Organics / BIONET

1,3-dimethyl 2-(hydroxyimino)propanedioate

  • MDL number MFCD00136215
  • Cas Number : 42937-74-6
  • Molecular Weight : 161.113

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70 - 72
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70805

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3G-908_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart