KEY001135264 Key Organics / BIONET

ethyl 5-methyl-4-[(1,3-thiazol-2-yl)carbamoyl]-1,2-oxazole-3-carboxylate

  • MDL number MFCD00664753
  • Cas Number : 338408-87-0
  • Molecular Weight : 281.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154 - 156
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70806

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3G-909_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart