KEY001135268 Key Organics / BIONET

3-(hydroxymethyl)-5-methyl-N-(1,3-thiazol-2-yl)-1,2-oxazole-4-carboxamide

  • MDL number MFCD00664755
  • Cas Number : 338408-93-8
  • Molecular Weight : 239.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230 - 232
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70810

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3G-913_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart