KEY001135311 Key Organics / BIONET

2-[(4-chlorophenyl)methoxy]ethan-1-ol

  • MDL number MFCD00172032
  • Cas Number : 1200-15-3
  • Molecular Weight : 186.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108 - 112@0.3mm Hg
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70855

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3G-963_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart