KEY001135406 Key Organics / BIONET

5-chloro-4-(2,4-dichlorophenyl)-1,2,3-thiadiazole

  • MDL number MFCD00173532
  • Cas Number : 338407-85-5
  • Molecular Weight : 265.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72 - 74
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70961

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3H-301S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart