KEY001135420 Key Organics / BIONET

1-(3,5-dichlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD00173546
  • Cas Number : 338408-13-2
  • Molecular Weight : 272.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 202 - 204
Iupac Name 1-(3,5-dichlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylic acid
Inchi Code 1S/C10H7Cl2N3O2/c1-5-13-9(10(16)17)14-15(5)8-3-6(11)2-7(12)4-8/h2-4H,1H3,(H,16,17)
Inchi Key CZTNDZALWLHXBA-UHFFFAOYSA-N

Description

Melting Point 202 - 204
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70978

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3H-323S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart