KEY001135465 Key Organics / BIONET

4-(2,4-dichlorophenyl)-1,2,3-thiadiazole-5-thiol

  • MDL number MFCD00173588
  • Cas Number : 66521-65-1
  • Molecular Weight : 263.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69 - 71
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71032

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3H-386S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart