KEY001135466 Key Organics / BIONET

4-(4-chlorophenyl)-1,2,3-thiadiazole-5-thiol

  • MDL number MFCD00173589
  • Cas Number : 338408-98-3
  • Molecular Weight : 228.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108 - 110
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71033

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3H-387S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart