KEY001135470 Key Organics / BIONET

2-{[(phenylcarbamoyl)methyl]sulfanyl}acetic acid

  • MDL number MFCD00202279
  • Cas Number : 70648-87-2
  • Molecular Weight : 225.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 106 - 108
Iupac Name [(2-anilino-2-oxoethyl)sulfanyl]acetic acid
Inchi Code 1S/C10H11NO3S/c12-9(6-15-7-10(13)14)11-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,12)(H,13,14)
Inchi Key ABHZTPDZBMDHPI-UHFFFAOYSA-N

Description

Melting Point 106 - 108
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71039

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3H-395S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart