KEY001135756 Key Organics / BIONET

1,3-thiazolidine-2,4-dicarboxylic acid

  • MDL number MFCD00145399
  • Cas Number : 30097-06-4
  • Molecular Weight : 177.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199 - 201
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71393

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3J-939_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart