KEY001135814 Key Organics / BIONET

[1-methyl-5-(4-methylphenoxy)-3-phenyl-1H-pyrazol-4-yl]methanol

  • MDL number MFCD00244906
  • Cas Number : 318234-01-4
  • Molecular Weight : 294.354

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150 - 152
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71477

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3K-366S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart