KEY001135857 Key Organics / BIONET

methyl 2-{[4-(hydroxymethyl)-1-methyl-3-phenyl-1H-pyrazol-5-yl]sulfanyl}benzoate

  • MDL number MFCD00244909
  • Cas Number : 318234-06-9
  • Molecular Weight : 354.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184 - 186
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71529

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3K-579S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart