KEY001135932 Key Organics / BIONET

2-[(4-chlorophenyl)sulfanyl]pyridine-3-carbaldehyde

  • MDL number MFCD00794341
  • Cas Number : 338982-28-8
  • Molecular Weight : 249.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86 - 88
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71614

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3L-303S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart