KEY001135933 Key Organics / BIONET

2-[(2,4-dichlorophenyl)sulfanyl]pyridine-3-carbaldehyde

  • MDL number MFCD00794342
  • Cas Number : 338982-29-9
  • Molecular Weight : 284.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114 - 117
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71615

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3L-304S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart