KEY001135937 Key Organics / BIONET

2-(4-methylphenoxy)pyridine-3-carbaldehyde

  • MDL number MFCD00794346
  • Cas Number : 478030-89-6
  • Molecular Weight : 213.236

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77 - 80
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71619

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3L-311S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart