KEY001136063 Key Organics / BIONET

5-chloro-1-methyl-3-[(phenylsulfanyl)methyl]-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00794869
  • Cas Number : 318234-26-3
  • Molecular Weight : 266.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105 - 107
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71747

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3L-578S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart