KEY001136112 Key Organics / BIONET

5-acetyl-1-(2-chloropyridin-3-yl)-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD00794645
  • Cas Number : 338413-91-5
  • Molecular Weight : 265.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224 - 226
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71799

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3L-917_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart