KEY001136181 Key Organics / BIONET

(2E)-1-[2-(4-chlorophenyl)-1,3-thiazol-5-yl]-3-(dimethylamino)prop-2-en-1-one

  • MDL number MFCD00974303
  • Cas Number : 338414-64-5
  • Molecular Weight : 292.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233 - 235
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71869

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3M-068_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart