KEY001136217 Key Organics / BIONET

4-oxo-1-(propan-2-yl)-1,4-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD01315709
  • Cas Number : 53977-47-2
  • Molecular Weight : 231.251

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200 - 203
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71911

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3M-352S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart