KEY001136354 Key Organics / BIONET

5-methyl-1,2-dihydro-1,6-naphthyridin-2-one

  • MDL number MFCD00867139
  • Cas Number : 88296-61-1
  • Molecular Weight : 160.176

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 246 - 248
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72057

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3N-012_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart